Learn More
Buerkle™ HDPE Lid for Round Tub
Buerkle™ HDPE Lid for Round Tub is suitable for use with Buerkle™ Tubs.
Brand:  Buerkle™ 3482-0751
38.74 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.89000kg
 View more versions of this product
Product Code. 10422372
  • 41.35 EUR / Each
Estimated Shipment: 24-07-2024
Description
  • Accessory for round tubs
  • Chemical resistance
  • Suitable for food
  • Made of HDPE
Specifications
Specifications
Lid
460 mm
1 Ea.
Polyethylene
For Round Tubs
-30°C to 70°C