Learn More
RETSCH Hardened Stainless Steel and Tungsten Carbide O-ring
Brand: RETSCH 05.114.0057
311.60 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.00300kg
Specifications
O-Ring | |
1Each | |
Hardened Stainless Steel and Tungsten Carbide | |
For Planetary Ball Mill PM 100 CM 50mL Grinding Jars |