Learn More
RETSCH Hardened Stainless Steel and Tungsten Carbide O-ring
Brand:  RETSCH 05.114.0056
185.25 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.00300kg
 View more versions of this product
Product Code. 10111842
  • 195.00 EUR / Each
Estimated Shipment: 31-07-2024
Specifications
O-Ring
1Each
Hardened Stainless Steel and Tungsten Carbide
For Planetary Ball Mill PM 100 CM 125mL Grinding Jars