Learn More
Cole-Parmer™ Hammond™ Desiccant
Choose indicating or non-indicating desiccant—indicating desiccant starts blue, then turns pink when regeneration or renewal is necessary. Use Hammond™; Drierite™ Desiccant (anhydrous calcium sulfate) in desiccators, drying columns or drying tubes.
Brand:  Cole-Parmer™ 07193-15
Additional Details : Weight : 2.20000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Chemical Name or Material Indicating Drierite Desiccant
Quantity 5 lb.
 View more versions of this product
Product Code. 11785589
Quantity Price
1 372.00 EUR / Pack of 4
6 338.52 EUR / Pack of 4
Estimated Shipment
23-04-2021
Description
Description
  • Indicating desiccant drying capacity is 10 to 14% of its weight for water and gases 
  • Non-indicating desiccant drying capacity is 6% of its weight for water and gases
Specifications
Chemical Identifiers
Specifications
Indicating Drierite Desiccant
Calcium Sulfate
5 lb.
Used in Desiccators, Drying Columns, or Drying Tubes
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Drierite, Indicating
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H400-Very toxic to aquatic life.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .