Learn More
Hamilton™ RN Adapters (Removable Adapters)
Adapters provide a connection to threaded fittings, tubing assemblies, tees, and crosses.
46.00 EUR - 52.00 EUR
Specifications
For Use With (Equipment) Removable Needle Syringe (250μL and Larger)
View More Specs
Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
11573144
View Documents Promotion Details
Hamilton™
35080
Male Luer-lok Hub Adapter
46.00 EUR
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
10041362
View Documents Promotion Details
Hamilton™
35081
Female Luer-lock Hub Adapter
52.00 EUR
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
Specifications
Removable Needle Syringe (250μL and Larger)