Learn More
Miele™ Half Insert
Brand: Miele™ 3808330
Additional Details : Weight : 50.00000kg
Specifications
Insert, Half | |
Flasks, graduated cylinder, bottles, etc., | |
186 mm | |
1 Each |
E 106/2 | |
420 mm | |
180 mm |