missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GST Rabbit, Europium, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
1088.00 EUR - 2785.00 EUR
Specifications
Antigen | GST |
---|---|
Classification | Polyclonal |
Conjugate | Europium |
Host Species | Rabbit |
Regulatory Status | RUO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15285684
|
Invitrogen
A15115 |
25 μg |
1088.00 EUR
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15295684
|
Invitrogen
A15116 |
100 μg |
2785.00 EUR
100µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GST | |
Europium | |
RUO | |
Primary |
Polyclonal | |
Rabbit | |
Drug Development | |
Store reagents at 2-8°C. |