Learn More
Deutsch Neumann™ Grey Rubber Spacers for Filtering (GUKO)
Excellent for general laboratory use. Deutsch Neumann™ Grey Rubber Spacers for Filtering (GUKO) are manufactured from natural rubber.
Brand: Deutsch Neumann™ 2010171
Additional Details : Weight : 0.12600kg
Description
- Durometer 40 Shore A
- Excellent cold flexibility
- High tensile strength and resilience
Specifications
Gray | |
1 Set | |
35 mm | |
58 mm |
Natural Rubber | |
Spacers | |
78 mm |