missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 69 K Smooth Surface Filter Discs Qualitative Filter Papers

Used for analytical and technical purposes.

Brand:  Sartorius™ FT-3-326-090

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11720476

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Used for analytical and technical purposes.
Specifications

Specifications

Disc
65μm/sec.
69 K
0.38 mm
90 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
50 Pack
Qualitative Filter Paper
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers