missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 69 K Smooth Surface Filter Discs Qualitative Filter Papers

Used for analytical and technical purposes.

Brand:  Sartorius™ FT-3-326-185

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11760476

  • 30.94 EUR / Pack of 50


Estimated Shipment
05-03-2021
Add to basket

Description

Description

Used for analytical and technical purposes.
Specifications

Specifications

50Pack
Qualitative
69 K
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
185mm
0.38mm
185mm
Refined Pulp and Linters
Filter discs
185mm
Videos
SDS
Documents