missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Grade 69 K Smooth Surface Filter Discs Qualitative Filter Papers
Used for analytical and technical purposes.
Brand: Sartorius™ FT-3-326-150
Additional Details : Weight : 0.01000kg
Description
Used for analytical and technical purposes.Specifications
50Pack | |
Qualitative | |
69 K | |
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications | |
150mm |
0.38mm | |
150mm | |
Refined Pulp and Linters | |
Filter discs | |
150mm |