missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 3 S/h Smooth Surface Qualitative Filter Papers

Used for routine analyses like clarification and determination of substances

Brand:  Sartorius™ FT-2-307-460570

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11745106

  • 257.00 EUR / Pack of 100
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

  • Made of refined pulp and cotton linters with an >95% α-cellulose content
  • Ash content between 0.1–0.15% (grade 100/N < 0.1%)
  • Wet-strengthened
Specifications

Specifications

Sheet
55μm/sec.
3 S/h
0.36 mm
460 L x 570 Wmm
570 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100 Pack
Qualitative Filter Paper
460 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers