Learn More
Fisherbrand™ Grade 122 General Purpose Filter Papers
A general purpose filter paper used for all general works in the laboratory for excellent and consistent filtration.
Brand:  Fisherbrand™
9.93 EUR valid until 2021-02-28
Use promo code "14802" to get your promotional price.
Additional Details : Weight : 0.16000kg
 View more versions of this product
Product Code. 11778742
Quantity Price
1 15.27 EUR / Pack of 100
10 14.07 EUR / Pack of 100
Estimated Shipment
12-03-2021
Add to basket
Description
Description
  • Very fast filtration velocity
  • Available in a range of sizes
  • Available as plain or folded discs
  • Applications include hospitals, research laboratories, schools, chemical industries, pharmaceuticals and more
Specifications
Specifications
100/pack
150mm
General works
Folded disc