Learn More
Fisherbrand™ Grade 122 Cellulose General Purpose Filter Paper
Very fast filtration velocity. Fisherbrand™ Grade 122 Cellulose General Purpose Filter Paper is used for all general works in laboratories.
Brand:  Fisherbrand™
Additional Details : Weight : 0.39600kg
 View more versions of this product
Product Code. 11788722
  • 31.90 EUR / Pack of 100
Estimated Shipment: 10-06-2024
Description
  • Very fast filtration velocity
  • Used for general works, hospitals, research laboratories
Applications:
  • Chemical industries, pharmaceutical, cosmetic, food and beverage
  • Specifications
    Specifications
    Folded Disc
    17 to 30 μm
    For all general works in laboratories
    100/Pk.
    250 mm
    Cellulose
    17 to 30 μm
    122
    0.17 mm
    250 mm