Learn More
Fisherbrand™ Grade 117 Special Filter Paper
Used for methylene blue test.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11750255
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Qualitatively indicates presence of hydroxyl radicals
  • Immediate, distinct bleaching of methylene blue dye on paper test strip
Application
  • Used to test for narrow-range pH solutions, where range is very low (0.2 to 2.0)
Specifications
Specifications
100PK
90mm
Paper used for Methylene blue test
90mm
4μm
117
Plain disc
0.2mm