Learn More
Fisherbrand™ Grade 115 Cellulose Very Slowly Filtering Qualitative Filter Paper
Fisherbrand™ Grade 115 Cellulose Very Slowly Filtering Qualitative Filter Paper are made of cellulose fibers.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
Product Code. 11475198
  • 26.65 EUR / Pack of 100
Estimated Shipment: 01-07-2024
Description
  • Pure filter paper grades made of cellulose fibers with alpha content of nearly 100% according precise norms
  • Ash content for these grades is approximatively 0.06 %