Learn More
Fisherbrand™ Grade 1103 Cellulose Technical Filter Paper
Suitable for removal of fine particles
Brand:  Fisherbrand™
Additional Details : Weight : 0.12150kg
 View more versions of this product
Product Code. 11739995
  • 10.15 EUR / Pack of 100
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Description
Description
  • Thick smooth paper
  • Wet strengthened
Specifications
Specifications
Disc
12 to 16 μm
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar
100 Pack
90 mm
Cellulose
12 to 16 μm
1103
0.3 mm
90 mm