Learn More
Fisherbrand™ Grade 1101 Cellulose Technical Filter Paper
Used for filtration of fatty products and suitable for hot filtrations.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11841713
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Thick creped paper
  • Wet strengthened
  • To filter syrups, essential oils, varnish, resinous solutions, paints, sweetened juices and wines
Specifications
Specifications
Disc
38 to 75 μm
For syrups, essential oils, varnish, resinous solutions, paints, sweetened juices, wines
100 Pack
185 mm
Cellulose
38 to 75 μm
1101
0.55 mm
185 mm