Learn More
Fisherbrand™ Grade 1101 Cellulose Technical Filter Paper
Used for filtration of fatty products and suitable for hot filtrations.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Thick creped paper
- Wet strengthened
- To filter syrups, essential oils, varnish, resinous solutions, paints, sweetened juices and wines
Spécification
Disc | |
38 to 75 μm | |
For syrups, essential oils, varnish, resinous solutions, paints, sweetened juices, wines | |
100 Pack | |
70 mm |
Cellulose | |
38 to 75 μm | |
1101 | |
0.55 mm | |
70 mm |