Learn More
Alfa Aesar™ Gold nanorods, 19-25nm dia. x 63-73nm length, dispersion in water, 700nm Absorption, negatively charged
Brand:  Alfa Aesar™ 046923.AA
Additional Details : CAS Number : 7440-57-5 Weight : 0.00005kg
Quantity 5mL
 View more versions of this product
Product Code. 15485045
  • 80.30 EUR / 5mL
Estimated Shipment date
from Supplier 31-08-2018
Add to basket
Specifications
Chemical Identifiers
7440-57-5
196.967
PCHJSUWPFVWCPO-UHFFFAOYSA-N
CHEBI:30050
Au
MFCD00003436
23985
Specifications
Gold nanorods
19 to 25nm dia. x 63 to 73nm length, dispersion in water, 700nm Absorption, negatively charged
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
[Au]
196.967
CHEBI:30050
Colorless
5mL
7440-57-5
MFCD00003436
Odorless
gold
23985
Liquid
>60°C (dec.)