missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Goat Thyroxine ELISA Kit (Colorimetric)

Product Code. 18214953 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214953 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214953 Supplier Novus Biologicals™ Supplier No. NBP2601571KIT
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Colorimetric ELISA Kit for study of thyroid function.

The Goat Thyroxine ELISA Kit (Colorimetric) measures goat Thyroxine in serum, plasma, tissue homogenates.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 20 to 320ng/mL
Assay Sensitivity 20ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum, Tissue Homogenates
Specificity For detection of goat T4
Target Species Goat
For Use With (Application) ELISA
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Signal Transduction
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym T4
Target Thyroxine
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.