Learn More
Goat anti-Chicken IgY (H+L) Secondary Antibody, DyLight 350, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75703
Product Code. 18240496
  • 277.00 EUR / 1mg
Description
IgY (H+L) Polyclonal specifically detects IgY (H+L) in Chicken samples. It is validated for Flow Cytometry, Immunomicroscopy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgY (H+L)
Polyclonal
DyLight 350
Goat
Affinity purified
RUO
Based on IEP, this antibody reacts with heavy gamma chains on chicken IgG/Y and light chains on all chicken immunoglobulins
Chicken
IgG
Flow Cytometry, Immunomicroscopy
LYOPH
Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Purified Chicken IgG/Y (H&L)
1 mg
Secondary
Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions