missing translation for 'onlineSavingsMsg'
Learn More

Glycol chitosan, 76.2%, MP Biomedicals™

Catalog Number p-4405296
Click to view available options
Quantity:
500 mg
1 g
5 g
This item is not returnable. View return policy
39280-86-9
(C8H15NO5)n
This item is not returnable. View return policy
39280-86-9
(C8H15NO5)n

Glycol Chitosan, Light Yellow Powder

  • Glycol chitosan increases membrane permeability and leakage in Glycine max Harosoy 63w cells.
Color Tan to White
Assay Percent Range 76.2%
Quantity 500 mg
Synonym Glycol chitin, deacetylated
Formula Weight (205.22)n
Physical Form Powder
Chemical Name or Material Glycol chitosan
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'