missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycol chitosan, 76.2%, MP Biomedicals™
Click to view available options
Quantity:
500 mg
1 g
5 g
Description
- Glycol chitosan increases membrane permeability and leakage in Glycine max Harosoy 63w cells.
Specifications
Color | Tan to White |
Assay Percent Range | 76.2% |
Quantity | 500 mg |
Synonym | Glycol chitin, deacetylated |
Formula Weight | (205.22)n |
Physical Form | Powder |
Chemical Name or Material | Glycol chitosan |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'