missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Solution, puriss p.a., 86 to 89% (T), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Poly bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Melting Point | 20°C |
Density | 1.252g/mL at 25°C |
Boiling Point | 290°C |
Flash Point | 160°C (320°F) |
Packaging | Poly bottle |
Linear Formula | HOCH2CH(OH)CH2OH · aq |
Refractive Index | 1.454 |
Quantity | 500 mL |
UN Number | NONH for all modes of transport |
Beilstein | 635685 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'