missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLS Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | GLS |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17561416
|
Proteintech
29519-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17551416
|
Proteintech
29519-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The GLS antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human GLS. This antibody recognizes human antigen. The GLS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
GLS | |
Polyclonal | |
Rabbit | |
2744 | |
Primary |
Western Blot | |
Unconjugated | |
Human | |
IgG |