Learn More
Mutexil™ Powder-Free Blue Nitrile Gloves
Catalog Number 15312316
Click to view available options
Size:
Large
Mutexil™ Powder-Free Blue Nitrile Gloves are disposable and come in a range of sizes. Tested for food contact.
  • Powder-free
  • Nitrile
  • Supplied in convenient dispenser boxes
  • Non-sterile
Certifications/Compliance AQL 1.5, EN 455-1-2&3, EN420, CE
Color Blue
Disposable Single-use
Hand Ambidextrous
Material Nitrile
Powdered Powder Free
Size Large
Sterility Non-sterile
Glove Cuff Style Beaded