Learn More
DWK Life Sciences Glass Stopper, Amber, SBW Glass, Standard Taper Ground Joint, Octagonal flat head
Amber SBW glass ground stopper.
Brand: DWK Life Sciences 246270701
Additional Details : Weight : 0.30000kg
Description
Solid amber SBW glass stoppers can be used to close glassware items such as flasks or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head.
- UV light protection due to external amber colouration
- Very durable and chemically resistant amber colour
- All glass components and therefore suitable for storage of corrosive and aggressive materials
- Tight closure due to standard taper ground joint
- Octagonal shaped head
Specifications
Storage of light sensitive reagents, pharmaceuticals, and chemicals; Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders | |
SBW Glass | |
No |
10/Pk. | |
ST 19/26 |