Learn More
DWK Life Sciences Glass Stopper, Amber, SBW Glass, Standard Taper Ground Joint, Octagonal flat head
Amber SBW glass ground stopper.
82.57 EUR - 113.00 EUR
Specifications
Autoclavable | No |
---|---|
Material | SBW Glass |
For Use With (Application) | Storage of light sensitive reagents, pharmaceuticals, and chemicals; Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders |
Quantity | 10/Pk. |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
15271989
|
DWK Life Sciences
246270607 |
ST 14/23 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15281989
|
DWK Life Sciences
246270701 |
ST 19/26 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Solid amber SBW glass stoppers can be used to close glassware items such as flasks or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head.
- UV light protection due to external amber colouration
- Very durable and chemically resistant amber colour
- All glass components and therefore suitable for storage of corrosive and aggressive materials
- Tight closure due to standard taper ground joint
- Octagonal shaped head
Specifications
No | |
SBW Glass | |
Storage of light sensitive reagents, pharmaceuticals, and chemicals; Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders | |
10/Pk. |