missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Hecht™ Assistent™ Wide Neck Screw Cap Bottles
Click to view available options
Capacity (Metric):
25 mL
100 mL
Description
- With thread and screw cap
- Clear glass
- Screw caps are heat resistant up to 110°C
Specifications
Closure Type | Screw Cap |
Product Type | Reagent Bottle |
Capacity (Metric) | 25 mL |
Diameter (Metric) Inner Neck | 28 mm |
Includes | Screw Cap |
Material | Clear Glass |
Volume (Metric) Dispense | 25mL |
Mouth | Wide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'