Learn More
Trajan Scientific and Medical™ Glass Lined Stainless-Steel GC Tubing
Exhibits resistance to the harshest of conditions and to strong acids and bases
Brand:  Trajan Scientific and Medical™ 082760
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15259003
  • 282.00 EUR / Each
Estimated Shipment: 03-06-2024
Description
  • Ideal for HPLC columns, particularly for biological matrices
  • Temperature stable up to 500°C
Specifications
Specifications
HPLC Columns, Biological Matrices
90 cm
Stable up to 500°C
2 mm
Borosilicate Glass (Lining)