Learn More
Merck Glass Fiber Filters with Binder, 2 μm
For prefiltration and contamination analysis
47.10 EUR - 191.00 EUR
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Filter Type | Depth Filter |
Material | Borosilicate Glass Microfiber |
Particle Retention | 0.8 to 8 μm |
Product Code | Brand | Diameter (English) | Grade | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Grade | Price | Quantity & Availability | ||||
10411751
|
Merck
AP2514250 |
5.59 in. | AP25 |
191.00 EUR
Pack of 50 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10677311
|
Merck
AP2501000 |
0.3 in. | AP25 |
47.10 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10771971
|
Merck
AP2502500 |
0.98 in. | AP25 |
52.20 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10586702
|
Merck
AP2009000 |
3.5 in. | AP20 |
121.00 EUR
Pack of 100 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
10712901
|
Merck
AP2012450 |
4.88 in. | AP20 |
163.00 EUR
Pack of 50 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10566512
|
Merck
AP2004700 |
1.8 in. | AP20 |
73.10 EUR
Pack of 100 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
10258330
|
Merck
AP2007500 |
2.9 in. | AP20 |
103.00 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10196370
|
Merck
AP2002500 |
0.98 in. | AP20 |
49.60 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10781971
|
Merck
AP2504200 |
1.6 in. | AP25 |
70.70 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10648451
|
Merck
AP2001300 |
0.5 in. | AP20 |
71.50 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10257960
|
Merck
AP2509000 |
3.5 in. | AP25 |
127.00 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10349840
|
Merck
AP2501300 |
0.5 in. | AP25 |
49.60 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10054660
|
Merck
AP2507500 |
2.9 in. | AP25 |
98.00 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10695571
|
Merck
AP2004200 |
1.6 in. | AP20 |
69.50 EUR
Pack of 100 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
10491361
|
Merck
AP2504700 |
1.8 in. | AP25 |
73.10 EUR
Pack of 100 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
10647891
|
Merck
AP2502200 |
0.8 in. | AP25 |
50.90 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10267960
|
Merck
AP2512450 |
4.88 in. | AP25 |
167.00 EUR
Pack of 50 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10722901
|
Merck
AP2014250 |
5.59 in. | AP20 |
187.00 EUR
Pack of 50 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10014640
|
Merck
AP2005500 |
2.1 in. | AP20 |
86.90 EUR
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
Description
For prefiltration and contamination analysis
Overview
Millipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121 °C at 1 bar).
Glass Fiber Filters with Binder Resin:Millipore filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated liquids. They are also widely used for clarification of aqueous solutions.Type AP20
- Lower retention, higher dirt-holding capacity than AP15 filters
- Recommended prefilter for 0.8 to 8.0 μm filters
- Use upstream to protect type AP15 filters
Type AP25
- Added thickness yields same retention and greater dirt-holding capacity as AP20
- Recommended prefilter for 0.9 to 8 μm filters, especially for proteinaceous and heavily contaminated liquids
- Use upstream to protect type AP15 filters
Specifications
Autoclavable | |
Depth Filter | |
0.8 to 8 μm | |
Sterile | |
Binder resin | |
1022°F | |
Filter Membranes |
White | |
Borosilicate Glass Microfiber | |
2 μm | |
Hydrophilic | |
Plain | |
550°C | |
Hydrophilic |