missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for VG-SYS Vari-gel MAXI Horizontal Electrophoresis Unit

For use with VG-SYS Vari-gel MAXI Horizontal Electrophoresis Unit.

Brand:  Fisherbrand™

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11756165

Quantity Price
1 71.83 EUR / Each
10 62.03 EUR / Each
Estimated Shipment
29-03-2021
Add to basket


Description

Description

  • Color coded
  • For use with VG-SYS Vari-gel MAXI Horizontal Electrophoresis Unit
Specifications

Specifications

VG-SYS Vari-Gel MAXI Horizontal Electrophoresis Unit
20
1 mm
5.9 mm
Comb
1 Each
29 μL
Videos
SDS
Documents