missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for HU10 Mini-Plus Horizontal Gel Electrophoresis Unit, 1.5mm thick

For use with HU10 Mini-Plus Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11759605

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Color coded
  • Height adjustable
  • Offer complete control over loading volume and well depth to maximum 32-sample throughput
Specifications

Specifications

HU10 Mini-Plus Horizontal Gel Electrophoresis Unit
20
20 μL
Comb
1.5 mm
3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers