Learn More
Biochrom SWIFT II Method System Control Software
Application modules for wavelength scanning, reaction kinetics, quantitation, multi-wavelength, time drive and fraction analysis
Brand: Biochrom 80-2108-31
Additional Details : Weight : 0.50000kg
Includes: Audit trail (data log) facility in each module
Description
- Print preview of results, with format customization
- Excellent export to spreadsheet capabilities
- Compatible with Ultrospec 2100, 3100, 3300 and 6300 pro
- Included as standard package for Ultrospec™ 4300 and 5300
Specifications
SWIFT II method - System control Software | |
Wavelength Scanning, Reaction Kinetics, Quantitation, Multi-Wavelength, Time Drive, Fraction Analysis, Tm and High Throughput Bacterial Cell Culture Work | |
Ultrospec Spectrophotometer | |
An audit trail (data log) facility in each module |