Learn More
Cytiva AutoSeq G-50 Columns
Use for rapid removal of unincorporated fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing. Cytiva Life Sciences™ illustra™ AutoSeq G-50 Columns provide effective purification essential for high-quality sequencing results.
196.00 EUR - 2199.00 EUR
Specifications
For Use With (Application) | Rapid removal of unincorporated Fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing |
---|---|
Product Type | Column |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11773309
|
Cytiva
27-5340-01 |
50 Columns |
196.00 EUR
Pack of 50 |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11783309
|
Cytiva
27-5340-02 |
250 Columns |
873.00 EUR
Pack of 250 |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11793309
|
Cytiva
27-5340-03 |
1000 Columns |
2199.00 EUR
Pack of 1000 |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Columns are prepacked with Sephadex G-50 DNA Grade F and pre-equilibrated in double-distilled water with 0.05% Kathon CG/ICP Biocide added as a preservative
- Columns that are ready to use enable sample application to collection of purified product in less than 4 min.
- Can be used to clean up reactions prepared with the BigDye Terminator v3.1 Cycle Sequencing Kit from Applied Biosystems
- Typical sample volume 12 to 25 μl
- Suitable for purification of DNA greater than 20 bases in length
- Optimized for automated fluorescent sequencing
Specifications
Rapid removal of unincorporated Fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing | |
Column |