missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Hybond™-N Membranes

Use for high sensitivity applications in DNA and RNA blotting. Cytiva Life Sciences™Amersham™ Hybond™-N Membranes is a strong supported membrane that is inherently hydrophilic and requires no pre-wetting.

Brand:  Cytiva RPN82N

Additional Details : Weight : 0.16356kg

 View more versions of this product

Product Code. 11778398

  • 155.00 EUR / Pack of 50


Estimated Shipment
15-03-2021
Add to basket

Description

Description

  • Binding capacity for nucleic acids up to 600μm/cm2
  • Inherently hydrophilic and requires no prewetting
  • Nucleic acids can be crosslinked using UV light

Specifications

Specifications

Disc
8.2cm
Up to 600μg/cm2
Nylon, Neutral
Southern and Northern blots; plaque lifts; UV fixation
82 mm
Videos
SDS
Documents