missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDPD1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-09605-100UL
Additional Details : Weight : 0.00970kg
Description
GDPD1 Polyclonal specifically detects GDPD1 in Rat samples. It is validated for Western Blot.Specifications
GDPD1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
EC 3.1, FLJ27503, FLJ37451, GDE4MGC35046, Glycerophosphodiester phosphodiesterase 4, glycerophosphodiester phosphodiesterase domain containing 1, glycerophosphodiester phosphodiesterase domain-containing protein 1 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat GDPD1 (NP_001037703). Peptide sequence MSSTAAFCLLSTLGGYLVTSFLLLKYPALLHQRKKQRFLSRHISHRGGAG | |
100 μg | |
Lipid and Metabolism | |
284161 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |