missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDI2 Mouse anti-Human, Mouse, Clone: 4B8F3, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | GDI2 |
---|---|
Clone | 4B8F3 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877045
|
Proteintech
60078-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867045
|
Proteintech
60078-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody can bind both GDIs for the close sequences.
Regulates the GDP/GTP exchange reaction of most Rab proteins by inhibiting the dissociation of GDP from them, and the subsequent binding of GTP to them.Specifications
GDI2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GDI 2, GDI2, GDP dissociation inhibitor 2, Rab GDI beta, RABGDIB | |
Gdi2 | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
4B8F3 | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Mouse, Human | |
P50395, Q61598 | |
14569, 2665 | |
GDI2 Fusion Protein Ag4613 | |
Primary | |
-20°C | |
GDI2 |