Learn More
GARP Monoclonal Antibody (YGIC86), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
160.00 EUR - 407.00 EUR
Specifications
Antigen | GARP |
---|---|
Clone | YGIC86 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15301840
|
Affymetrix eBioscience
48-9891-82 |
100 μg |
407.00 EUR
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15391830
|
Affymetrix eBioscience
48-9891-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The YGIC86 monoclonal antibody reacts with mouse Glycoprotein A Repetitions Predomit (GARP, also known as Lrrc32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on Treg cells has been reported to be necessary for the suppressive function of Treg cells, possibly related to its role as a cell surface receptor for LAP/TGF beta. When looking for expression of GARP on platelets, it is recommended to use the Foxp3 Staining Buffer Set (cat. 00-5523) to fix the platelets before staining with this YGIC86 monoclonal antibody. Fixation/permeabilization is not necessary when staining Treg cells. YGIC86 does not cross-react to rat GARP. Applications Reported: This YGIC86 antibody has been reported for use in flow cytometric analysis. Applications Tested: This YGIC86 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
GARP Monoclonal antibody specifically detects GARP in Mouse samples. It is validated for Flow Cytometry.Specifications
GARP | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
434215 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
YGIC86 | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.