Learn More
Gardner (ASTM D1544, D6166) Colour Reference Standard, Paragon Scientific™
Gardner Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
Brand:  Paragon Scientific 134200
Additional Details : CAS Number : 8042-47-5 Weight : 0.90000kg
 View more versions of this product
Product Code. 16020162
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to International Standards
  • 500mL Pack Size supplied with MSDS
  • 18 months shelf life
  • Tamper evident security packaging
  • Manufactured in the United kingdom
Specifications
Specifications
500 mL
Color Reference Standard
Special Offers
Nominal values quoted