missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOS Mouse anti-Human, Mouse, Clone: 1G2C5, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | FOS |
---|---|
Clone | 1G2C5 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16868605
|
Proteintech
66590-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16858605
|
Proteintech
66590-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FBJ murine osteosarcoma viral oncogene homolog is commonly known as Cellular protooncogene Fos (c-Fos). It belongs to the fos transcription factor family and is encoded by the C-FOS gene located on chromosome 14. c-Fos heterodimerizes with c-Jun to form the Activator Protein -1 (AP-1) transcription factor which regulates transcription of several genes that play an important role in cellular signal transduction, cell proliferation and differentiation. c-Fos has been shown to be phosphorylated by various stimuli including growth factors and insulin at atleast seven different sites. The best known upstream kinase for phosphorylation at Threonine 232 is ERK MAPK which regulates localization of c-Fos to the nucleus and is thus important for c-Fos induced transcriptional activity.Specifications
FOS | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AP 1, C FOS, Cellular oncogene fos, FOS, G0S7, Proto oncogene c Fos | |
FOS | |
IgG1 | |
Protein G | |
Antibody |
1G2C5 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P01100, P01101 | |
14281, 2353 | |
FOS Fusion Protein Ag24340 | |
Primary | |
-20°C | |
FOS |