missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic acid, Reagent grade, Honeywell
Click to view available options
Quantity:
100 mL
500 mL
1 L
Specifications
Linear Formula | HCOOH |
UN Number | UN1779 |
Beilstein | 1209246 |
Formula Weight | 46.03g/mol |
Density | 1.22g/mL at 25°C |
Percent Purity | ≥95% |
Grade | Reagent |
Quantity | 1 L |
Explosion Limit | 0.57 |
Flash Point | 49°C (120.2°F) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'