missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, 85% w/w, Extra Pure, Scharlab
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
HDPE Carboy
Description
Extra Pure grade specifications are less strict than Reagent Grade, but are significantly higher than industrial products. They are often used in the pharmaceutical industry, either as a reagent or as raw materials for production.
Specifications
Melting Point | -9°C |
Boiling Point | 107°C |
Color | Colorless |
Linear Formula | HCOOH |
UN Number | 3412 |
Solubility Information | Miscible in water |
Formula Weight | 46.03g/mol |
Percent Purity | 85% (min.) |
Grade | Applications: analytical chemistry, synthesis of organic products, in the rubber industry, acidifying agent, cosmetics |
Density | 1.2g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'