missing translation for 'onlineSavingsMsg'
Learn More

Follistatin Antibody (17-2), DyLight 680, Novus Biologicals™

Product Code. 18214721 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214721 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214721 Supplier Novus Biologicals Supplier No. NBP261941FR
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Follistatin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 17-2
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias follistatin, follistatin isoform FST317, FSActivin-binding protein
Gene Symbols FST
Host Species Mouse
Immunogen Recognizes human Follistatin isoform FS288, raised against recombinant human FS288
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10468
Test Specificity This antibody recognizes human Follistatin isoform FS288.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.