Learn More
BioTek Instruments Fluorescence Filter
Fluorescence Filter for use with BioTek Microplate readers and imagers: Cytation 5, Synergy H1, Synergy HTX and Synergy LX
Brand:  BioTek Instruments 7082213
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10003824
  • 422.00 EUR / Each
Estimated Shipment: 21-01-2022
Description
Description
Fluorescence Filter for use with BioTek Microplate readers and imagers: Cytation 5, Synergy H1, Synergy HTX and Synergy LX
Specifications
Specifications
620/40 nm
Cytation 3, Cytation 5, FLx800, Synergy 2, Synergy H1, Synergy HTX Multi-Mode Readers
Fluorescence Filter