missing translation for 'onlineSavingsMsg'
Learn More

FLT3 [D835Y] Recombinant Human Protein, Invitrogen™

FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family.

Brand:  Invitrogen PR7450A

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15458593

  • 2150.00 EUR / 100µg


Estimated Shipment date
from Supplier 25-01-2019
Add to basket

Description

Description

The FLT3 D835Y mutant is a drug resistant form of FLT3.

Specifications

Specifications

NP_004110, D835Y
Frozen
48.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
PDGFR Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 100mM NaCl, 0.5mM EDTA, 0.05% NP-40, 2mM DTT, 50% glycerol
100μg
RUO - research use only
FLT3 D835Y, FLK2, STK1, CD135
Baculovirus
Human
His Tag
Videos