Learn More
Cytiva Flow Cell
UV flow cell
Brand: Cytiva 18-1128-25
Additional Details : Weight : 0.03200kg
Description
UV Flow CellSpecifications
Flow Cell | |
2 μL | |
ETFE, Glass, Titanium, Fluorocarbon Rubber O-ring | |
2 mm |