missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimble Chase™ Flex-Column™ Borosilicate Glass Economy Column
Brand: Kimble Chase™ 4204001511
Additional Details : Weight : 0.80000kg
Specifications
Ion exchange chromatography | |
0 to 50°C | |
Economy Column | |
1 bar | |
1000 mm | |
1.77cm3 |
3 Pack | |
15 mm | |
Type I | |
177 mL | |
Borosilicate Glass |