missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flavin Adenine Dinucleotide Disodium Salt, ≥95%, MP Biomedicals™
Click to view available options
Quantity:
250 mg
500 mg
1 g
Description
Cell Biology and Analysis
Specifications
Color | Orange |
Assay Percent Range | ≥95% |
Quantity | 250 mg |
IUPAC Name | disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate |
Formula Weight | 829.6 |
Physical Form | Solid |
Chemical Name or Material | Flavin Adenine Dinucleotide Disodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'